missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Dinitro-2-methyl-d3-phenol, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-4,6-dinitrophenol methyl-d3 |
| CAS | 1219804-69-9 |
| Molecular Formula | C7 2H3 H3 N2 O5 |
| Quantity | 0.05 g |
| Synonym | DNOC D3 (methyl-D3), 2-Methyl-4,6-dinitrophenol (methyl-D3), 2-Trideuteriomethyl-4,6-dinitrophenol, 4,6-Dinitro-2-trideuteriomethylphenol |
| InChI Formula | InChI=1 S/C7H6N2O5/c1-4-2-5(8(11)12)3-6(7(4)10)9(13)14/h2-3,10 H,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])c1cc(cc(c1O)[N+](=O)[O-])[N+](=O)[O-] |
| IUPAC Name | 2,4-dinitro-6-(trideuteriomethyl)phenol |
| Molecular Weight (g/mol) | 201.15 |
| Formula Weight | 201.0465 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction