missing translation for 'onlineSavingsMsg'
Learn More

4-[(6-Methylpyrazin-2-yl)oxy]benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10347444
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10347444 250 mg Amber Glass Bottle 250mg
10718265 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10347444 Supplier Thermo Scientific Maybridge Supplier No. CC62703CB
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 926921-66-6
Molecular Formula C11H9ClN2O3S
Molecular Weight (g/mol) 284.71
MDL Number MFCD09879936
InChI Key GUJMBSPJUZJPEB-UHFFFAOYSA-N
Synonym 4-6-methylpyrazin-2-yl oxy benzenesulfonyl chloride, 4-6-methylpyrazin-2-yl oxy benzene-1-sulfonyl chloride
PubChem CID 24229688
IUPAC Name 4-(6-methylpyrazin-2-yl)oxybenzenesulfonyl chloride
SMILES CC1=CN=CC(OC2=CC=C(C=C2)S(Cl)(=O)=O)=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 284.72
Percent Purity 97%
Chemical Name or Material 4-[(6-Methylpyrazin-2-yl)oxy]benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.