missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4,6-O-Ethylidene-D-glucopyranose, 90%

Product Code. 15497989
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497989 1 g 1g
15407999 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497989 Supplier Thermo Scientific Chemicals Supplier No. J66583.03
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13224-99-2 | C8H14O6 | 206.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13224-99-2
Molecular Formula C8H14O6
Molecular Weight (g/mol) 206.19
MDL Number MFCD00006820
InChI Key VZPBLPQAMPVTFO-UHFFFAOYNA-N
Synonym 4,6-o-ethylidene-a-d-glucose
PubChem CID 21581900
IUPAC Name (2R,4aR,6S,7R,8R,8aS)-2-methyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxine-6,7,8-triol
SMILES CC1OCC2OC(O)C(O)C(O)C2O1

Specifications

Color White
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 206.19
Percent Purity 90%
Physical Form Powder
Chemical Name or Material 4,6-O-Ethylidene-D-glucopyranose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.