Learn More
4,7-Dibromo-2,1,3-benzothiadiazole, 97%
Description
4,7-Dibromo-2,1,3-benzothiadiazole is the building block or monomer for the synthesis of light-emitting diodes and conducting polymers for organic electronics. It is used as an intermediate for the synthesis of poly[N-9'-heptadecanyl-2,7-carbazole-alt-5,5-(4',7'-di-2-thienyl-2',1',3'-benzothiadiazole)] (PCDTBT) and poly[2,1,3-benzothiadiazole-4,7-diyl[4,4-bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene-2,6-diyl]] (PCPDTBT).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4,7-Dibromo-2,1,3-benzothiadiazole is the building block or monomer for the synthesis of light-emitting diodes and conducting polymers for organic electronics. It is used as an intermediate for the synthesis of poly[N-9′-heptadecanyl-2,7-carbazole-alt-5,5-(4′,7′-di-2-thienyl-2′,1′,3′-benzothiadiazole)] (PCDTBT) and poly[2,1,3-benzothiadiazole-4,7-diyl[4,4-bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b′]dithiophene-2,6-diyl]] (PCPDTBT).
Solubility
Soluble in toluene.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 186°C to 190°C |
| Quantity | 25 g |
| UN Number | UN2811 |
| Solubility Information | Soluble in toluene. |
| Formula Weight | 293.97 |
| Percent Purity | 97% |
| Chemical Name or Material | 4,7-Dibromo-2,1,3-benzothiadiazole |
Safety and Handling
- 4,7-Dibromo-2,1,3-benzothiadiazole
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : 4,7-Dibromobenzo[c]-1,2,5-thiadiazole
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.