missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dihydroxy-1,10-phenanthroline

Product Code. 11327468
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327468 2 g 2g
11337468 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327468 Supplier Thermo Scientific Alfa Aesar Supplier No. 031434.04
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3922-40-5 | C12H8N2O2 | 212.208 g/mol

4,7-Dihydroxy-1,10-phenanthroline is used as a phenanthroline stain. Its iron(II) complex reacts rapidly with dissolved oxygen in aqueous solution, thereby it is utilized for the spectrophotometric determination of dissolved oxygen up to 20 ppm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3922-40-5
Molecular Formula C12H8N2O2
Molecular Weight (g/mol) 212.208
MDL Number MFCD00004975
InChI Key SLIBCJURSADKPV-UHFFFAOYSA-N
Synonym 1,10-phenanthroline-4,7-diol, 4,7-dihydroxy-1,10-phenanthroline, 1,10-phenanthroline-4,7 1h,10h-dione, 1,4,7,10-tetrahydro-1,10-phenanthroline-4,7-dione, pyridino 3,2-h quinoline-4,7-diol, acmc-1coeh, bidd:gt0532, 1,1-phenanthroline-4,7-diol, 1,10 phenanthroline-4,7-diol, 4,7-dioxylato-1,10-phenanthroline
PubChem CID 77524
IUPAC Name 1,10-dihydro-1,10-phenanthroline-4,7-dione
SMILES C1=CC2=C(C3=C1C(=O)C=CN3)NC=CC2=O

Specifications

Melting Point >300°C (decomposition)
Color Yellow
Quantity 2 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 212.21
Physical Form Powder
Chemical Name or Material 4,7-Dihydroxy-1,10-phenanthroline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.