missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dimethoxy-1,10-phenanthroline, 97%

Product Code. 10763342 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10763342 250 mg Glass bottle 250mg
10057999 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10763342 Supplier Thermo Scientific Acros Supplier No. 433782500
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92149-07-0 | C14H12N2O2 | 240.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92149-07-0
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
MDL Number MFCD00233883
InChI Key ZPGVCQYKXIQWTP-UHFFFAOYSA-N
Synonym 4,7-dimethoxy-1,10-phenanthroline, acmc-20a0a1, 4,7-dimethoxy-1,10-phenthroline, 4?7-dimethoxy-1?10-phenanthroline, 1,10-phenanthroline, 4,7-dimethoxy, 4,7-bis methoxy-1,10-phenanthroline, 4,7-bis methyloxy-1,10-phenanthroline
PubChem CID 10462298
IUPAC Name 4,7-dimethoxy-1,10-phenanthroline
SMILES COC1=C2C=CC3=C(C=CN=C3C2=NC=C1)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 212.0°C to 214.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 240.26
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dimethoxy-1,10-phenanthroline
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4,7-Dimethoxy-1,10-phenanthroline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.