missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dimethylpyrazolo[5,1-c][1,2,4]triazine-3-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10374092
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10374092 10 g Amber Glass Bottle 10g
10252832 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10374092 Supplier Thermo Scientific Maybridge Supplier No. KM01115EA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175137-58-3
Molecular Formula C8H8N4O2
Molecular Weight (g/mol) 192.178
MDL Number MFCD00067873
InChI Key JFCXJLMOHOXSTO-UHFFFAOYSA-N
Synonym 4,7-dimethylpyrazolo 5,1-c 1,2,4 triazine-3-carboxylic acid, 4,7-dimethylpyrazolo 3,2-c 1,2,4 triazine-3-carboxylic acid, 3-carboxy-4,7-dimethylpyrazolo 5,1-c 1,2,4 triazine, maybridge1_004683, 4,7-dimethylpyrazolo 5,1-c 1,2,4-triazine-3-carboxylic acid, 4,7-dimethylpyrazolo 5,1-c triazine-3-carboxylic acid, 4,7dimethyl-pyrazolo 5,1-c 1,2,4 triazine-3-carboxylic acid, pyrazolo 5,1-c 1,2,4 triazine-3-carboxylic acid, 4,7-dimethyl, pyrazolo 5,1-c 1,2,4 triazine-3-carboxylicacid, 4,7-dimethyl, pyrazolo 5,1-c 1,2,4 triazine-3-carboxylicacid,4,7-dimethyl
PubChem CID 2778429
IUPAC Name 4,7-dimethylpyrazolo[5,1-c][1,2,4]triazine-3-carboxylic acid
SMILES CC1=NN2C(=C(N=NC2=C1)C(=O)O)C

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 192.18
Percent Purity 95%
Chemical Name or Material 4,7-Dimethylpyrazolo[5,1-c][1,2,4]triazine-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.