missing translation for 'onlineSavingsMsg'
Learn More

4-(9-Carbazolyl)benzeneboronic acid, 98%

Product Code. 15472009
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15472009 1 g 1g
15492009 250 mg 250mg
15482009 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15472009 Supplier Thermo Scientific Alfa Aesar Supplier No. H64703.03
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 419536-33-7 | C18H14BNO2 | 287.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 419536-33-7
Molecular Formula C18H14BNO2
Molecular Weight (g/mol) 287.13
MDL Number MFCD13176534
InChI Key JGAVTCVHDMOQTJ-UHFFFAOYSA-N
Synonym 4-9h-carbazol-9-yl phenyl boronic acid, 4-9h-carbozol-9-yl phenylboronic acid, 4-9h-carbazol-9-yl phenylboronic acid, 4-9-carbazolyl benzeneboronic acid, 4-9h-9-carbozale phenylboronic acid, 4-9-carbazolyl phenylboronic acid, 4-carbazol-9-yl phenylboronic acid, boronic acid, 4-9h-carbazol-9-yl phenyl, amtb338
PubChem CID 23120827
SMILES OB(O)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White
Quantity 1 g
Beilstein 10565612
IUPAC Name (4-carbazol-9-ylphenyl)boronic acid
Formula Weight 287.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-(9-Carbazolyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.