missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 4-Acetamido-4'-Maleimidylstilbene-2,2'-Disulfonic Acid, Disodium Salt

Product Code. 11554107
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11554107 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11554107 Supplier Invitrogen™ Supplier No. A485
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

4-acetamido-4'-maleimidylstilbene-2,2'-disulfonic acid, disodium salt

Our 4-acetamido-4'-maleimidylstilbene-2,2'- disulfonic acid is a thiol-reactive reagent that is water soluble, with high polarity and membrane impermeability. This polysulfonated dye is useful for determining whether thiol-containing proteins and polypeptide chains are exposed at the extracellular or cytoplasmic membrane surface.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Product Type 4-Acetamido-4'-Maleimidylstilbene-2,2'-Disulfonic Acid
Content And Storage Store in freezer (-5 to -30°C).
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Maleimide
Label or Dye Stilbene
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.