missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidoacetophenone, 98%

Product Code. 10238753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10238753 5 g Glass bottle 5g
10025343 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10238753 Supplier Thermo Scientific Acros Supplier No. 391820050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2719-21-3 | C10H11NO2 | 177.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2719-21-3
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.2
MDL Number MFCD00014965
InChI Key WECHHDJTILFYQT-UHFFFAOYSA-N
Synonym 4'-acetamidoacetophenone, n-4-acetylphenyl acetamide, 4-acetamidoacetophenone, n-p-acetylphenyl acetamide, acetamide, n-4-acetylphenyl, p-acetylaminoacetophenone, 4-acetylacetanilide, 4'-acetylacetanilide, p-acetamidoacetophenone, 4-acetamido-acetophenone
PubChem CID 75937
IUPAC Name N-(4-acetylphenyl)acetamide
SMILES CC(=O)C1=CC=C(C=C1)NC(=O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 166°C to 170°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3COC6H4NHCOCH3
Quantity 5 g
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in benzene
Formula Weight 177.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamidoacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 4-Acetamidoacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.