missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidophenol, 98%, Thermo Scientific™

42.20 € - 420.00 €

Chemical Identifiers

CAS 103-90-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00002328
InChI Key RZVAJINKPMORJF-UHFFFAOYSA-N
Synonym acetaminophen, 4-acetamidophenol, paracetamol, apap, n-4-hydroxyphenyl acetamide, tylenol, acetaminofen, panadol, datril, p-acetamidophenol
PubChem CID 1983
ChEBI CHEBI:46195
IUPAC Name N-(4-hydroxyphenyl)acetamide
SMILES CC(=O)NC1=CC=C(O)C=C1
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11473574
SDS View Documents
Thermo Scientific Alfa Aesar
A11240.30
250g
42.20 €
250g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
11359439
SDS View Documents
Thermo Scientific Alfa Aesar
A11240.36
500g
67.80 €
500g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
11463574
SDS View Documents
Thermo Scientific Alfa Aesar
A11240.0B
1000g
112.00 €
1000g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
11483574
SDS View Documents
Thermo Scientific Alfa Aesar
A11240.0I
5000g
420.00 €
5000g
View alternative products
Save Up to 
»
Estimated Shipment: 06-10-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Cyclooxygenase inhibitor and widely used analgesic. 4-Acetamidophenol formulated pharmaceutical products are used as antiinfectant, analgesic, anti rheumatic and antipyretic. It is used as an intermediate in organic synthesis, hydrogen peroxide stabilizer and photographic chemicals.

Solubility
Soluble in alcohol, dimethylformamide, ethylene dichloride, acetone and ethyl acetate. Slightly soluble in water and ether.

Notes
Incompatible with strong oxidizing agents, acids and alkali metals.
Specifications

Chemical Identifiers

103-90-2
151.17
RZVAJINKPMORJF-UHFFFAOYSA-N
1983
N-(4-hydroxyphenyl)acetamide
C8H9NO2
MFCD00002328
acetaminophen, 4-acetamidophenol, paracetamol, apap, n-4-hydroxyphenyl acetamide, tylenol, acetaminofen, panadol, datril, p-acetamidophenol
CHEBI:46195
CC(=O)NC1=CC=C(O)C=C1

Specifications

103-90-2
C8H9NO2
2208089
acetaminophen, 4-acetamidophenol, paracetamol, apap, n-4-hydroxyphenyl acetamide, tylenol, acetaminofen, panadol, datril, p-acetamidophenol
RZVAJINKPMORJF-UHFFFAOYSA-N
N-(4-hydroxyphenyl)acetamide
1983
151.17
4-Acetamidophenol
1.293
MFCD00002328
14,47
Soluble in alcohol,dimethylformamide,ethylene dichloride,acetone and ethyl acetate. Slightly soluble in water and ether.
CC(=O)NC1=CC=C(O)C=C1
151.17
CHEBI:46195
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 203-157-5

missing translation for 'rtecsNumber' : AE4200000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • 4-Acetamidophenol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contain: Acetaminophen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.