missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidophenol, 98%

Product Code. 11483574
Change view
Click to view available options
Quantity:
250 g
500 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483574 5000 g 5000g
11473574 250 g 250g
11359439 500 g 500g
11463574 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11483574 Supplier Thermo Scientific Alfa Aesar Supplier No. A11240.0I
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103-90-2 | C8H9NO2 | 151.17 g/mol

Cyclooxygenase inhibitor and widely used analgesic. 4-Acetamidophenol formulated pharmaceutical products are used as antiinfectant, analgesic, anti rheumatic and antipyretic. It is used as an intermediate in organic synthesis, hydrogen peroxide stabilizer and photographic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclooxygenase inhibitor and widely used analgesic. 4-Acetamidophenol formulated pharmaceutical products are used as antiinfectant, analgesic, anti rheumatic and antipyretic. It is used as an intermediate in organic synthesis, hydrogen peroxide stabilizer and photographic chemicals.

Solubility
Soluble in alcohol, dimethylformamide, ethylene dichloride, acetone and ethyl acetate. Slightly soluble in water and ether.

Notes
Incompatible with strong oxidizing agents, acids and alkali metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-90-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00002328
InChI Key RZVAJINKPMORJF-UHFFFAOYSA-N
Synonym acetaminophen, 4-acetamidophenol, paracetamol, apap, n-4-hydroxyphenyl acetamide, tylenol, acetaminofen, panadol, datril, p-acetamidophenol
PubChem CID 1983
ChEBI CHEBI:46195
IUPAC Name N-(4-hydroxyphenyl)acetamide
SMILES CC(=O)NC1=CC=C(O)C=C1

Specifications

Melting Point 168°C to 172°C
Density 1.293
Quantity 5000 g
Beilstein 2208089
Merck Index 14,47
Solubility Information Soluble in alcohol,dimethylformamide,ethylene dichloride,acetone and ethyl acetate. Slightly soluble in water and ether.
Formula Weight 151.17
Percent Purity 98%
Chemical Name or Material 4-Acetamidophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.