missing translation for 'onlineSavingsMsg'
Learn More

4-Acetaminophen Sulfate Potassium Salt, TRC

Product Code. 30133197
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133197 25 mg 25mg
17901425 5 mg 5mg
17973355 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133197 Supplier Toronto Research Chemicals Supplier No. TRCA16123025MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32113-41-0
Molecular Formula C8 H8 N O5 S . K
Molecular Weight (g/mol) 269.32
Synonym Acetamide, N-[4-(sulfooxy)phenyl]-, potassium salt (1:1) (ACI), Acetamide, N-[4-(sulfooxy)phenyl]-, monopotassium salt (9CI), Acetanilide, 4'-hydroxy-, hydrogen sulfate (ester), monopotassium salt (8CI), Potassium N-acetyl-p-aminophenyl sulfate, N-Acetyl-p-aminophenyl sulfate potassium salt, Paracetamol O-sulfate potassium salt, Paracetamol sulfate potassium, Potassium paracetamol O-sulfate, Paracetamol O-sulfate potassium
IUPAC Name potassium;(4-acetamidophenyl) sulfate
SMILES [K+].CC(=O)Nc1ccc(OS(=O)(=O)[O-])cc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C8H9NO5S.K/c1-6(10)9-7-2-4-8(5-3-7)14-15(11,12)13;/h2-5H,1H3,(H,9,10)(H,11,12,13);/q;+1/p-1
Formula Weight 268.976
Percent Purity >95%
Chemical Name or Material 4-Acetaminophen Sulfate Potassium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.