missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxy-N-isopropyl-N-methyltryptamine, TRC

Product Code. 30323945
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323945 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323945 Supplier Toronto Research Chemicals Supplier No. TRCA1656255MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Acetoxy-N-isopropyl-N-methyltryptamine
CAS 1024612-25-6
Molecular Formula C16H22N2O2
Quantity 5 mg
Synonym 4-Acetate 3-[2-[Methyl(1-methylethyl)amino]ethyl]-1H-indol-4-ol
InChI Formula InChI=1S/C16H22N2O2/c1-11(2)18(4)9-8-13-10-17-14-6-5-7-15(16(13)14)20-12(3)19/h5-7,10-11,17H,8-9H2,1-4H3
SMILES CC(C)N(C)CCc1c[nH]c2cccc(OC(=O)C)c12
IUPAC Name [3-[2-[methyl(propan-2-yl)amino]ethyl]-1H-indol-4-yl] acetate
Molecular Weight (g/mol) 274.36
Formula Weight 274.1681
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.