missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbenzoic acid, 98%

Product Code. 10163760 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10163760 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10163760 Supplier Thermo Scientific Acros Supplier No. 175860100
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 586-89-0 | C9H8O3 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 586-89-0
CAS Min % 97.5
CAS Max % 100.0
Melting Point 206.0°C to 210.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C9H8O3
Linear Formula CH3COC6H4CO2H
MDL Number MFCD00002561
Quantity 10 g
Synonym benzoic acid, 4-acetyl, p-acetylbenzoic acid, 4-acetyl-benzoic acid, 4'-acetophenonecarboxylic acid, 4-acetylbezoic acid, 4-acetylbenzoci acid, 4'-carboxyacetohenone, pubchem3129, 4-acetyl benzoic acid
Solubility Information Solubility in water: soluble.
InChI Key QBHDSQZASIBAAI-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=C(C=C1)C(=O)O
IUPAC Name 4-acetylbenzoic acid
Molecular Weight (g/mol) 164.16
PubChem CID 11470
Formula Weight 164.16
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Acetylbenzoic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Acetylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.