missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylphenyl Beta-D-Glucopyranoside, TRC

Product Code. 30009986
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009986 50 mg 50mg
30004000 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009986 Supplier Toronto Research Chemicals Supplier No. TRCA18739050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-14-3
Molecular Formula C14 H18 O7
Molecular Weight (g/mol) 298.29
Synonym Ethanone, 1-[4-(β-D-glucopyranosyloxy)phenyl]-, Picein (6CI,7CI,8CI), 1-[4-(β-D-Glucopyranosyloxy)phenyl]ethanone, 4-Acetylphenyl β-D-glucopyranoside, Ameliaroside, Piceoside, Salicinerein, Salinigrin, p-Hydroxyacetophenone glucoside, p-Hydroxyacetophenone-D-glucoside
IUPAC Name 1-[4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]ethanone
SMILES CC(=O)c1ccc(O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)cc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H18O7/c1-7(16)8-2-4-9(5-3-8)20-14-13(19)12(18)11(17)10(6-15)21-14/h2-5,10-15,17-19H,6H2,1H3/t10-,11-,12+,13-,14-/m1/s1
Formula Weight 298.1053
Percent Purity >95%
Chemical Name or Material Picein
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.