missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylphenylboronic acid pinacol ester, 97%

Product Code. 11956031 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11956031

Brand: Thermo Scientific Acros 444430010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 171364-81-1 | C14H19BO3 | 246.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 171364-81-1
Molecular Formula C14H19BO3
Molecular Weight (g/mol) 246.11
MDL Number MFCD05863922
InChI Key BATKIZWNRQGSKE-UHFFFAOYSA-N
Synonym 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-acetylphenylboronic acid pinacol ester, 4-acetylphenylboronic acid, pinacol ester, 2-4-acetylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethan-1-one, amtb682, 4-acetylphenyl boronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2yl phenyl-ethanone
PubChem CID 2760596
IUPAC Name 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 59°C to 62°C
Color White
Boiling Point 80°C (0.1 Torr)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 246.12
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetylphenylboronic acid pinacol ester
Show More Show Less
compliance-icons
Product Identifier
  • 4-Acetylphenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.