missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1,2,2,6,6-pentamethylpiperidine, 99%

Product Code. 10546451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10546451 1 g Glass bottle 1g
10285911 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10546451 Supplier Thermo Scientific Acros Supplier No. 335940010
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 40327-96-6 | C10H24N2 | 172.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40327-96-6
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.32
MDL Number MFCD01861832
InChI Key CGXOAAMIQPDTPE-UHFFFAOYSA-P
Synonym 4-amino-1,2,2,6,6-pentamethylpiperidine, 1,2,2,6,6-pentamethyl-4-aminopiperidine, 1,2,2,6,6-pentamethyl-4-piperidinamine, 1,2,2,6,6-pentamethyl-4-piperidylamine, 4-ammo-1,2,2,6,6-pentamethylpiperidine, 4-piperidinamine,1,2,2,6,6-pentamethyl, 1,2,-2,6,6-pentamethyl-4-aminopiperidine, 1,2,2,6,6,-pentamethyl-4-aminopiperidine, 4-amino-1,2,2,6,6-pentamethyl-piperidine, 4-amino-1,2,2,6,6-pentamethyl piperidine, 97+%
PubChem CID 693792
IUPAC Name 1,2,2,6,6-pentamethylpiperidin-4-amine
SMILES C[NH+]1C(C)(C)CC([NH3+])CC1(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 215°C to 218°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4795 to 1.4815
Quantity 1 g
Beilstein 22, II, 321
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents
Formula Weight 170.3
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Amino-1,2,2,6,6-pentamethylpiperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.