missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1,2,4-triazole, 99%

Product Code. 11433308
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433308 100 g 100g
11413308 5 g 5g
11423308 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433308 Supplier Thermo Scientific Alfa Aesar Supplier No. A17716.22
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 584-13-4 | C2H4N4 | 84.082 g/mol

4-Amino-1,2,4-triazole is used as an intermediate for the preparation of antifungal agent like fluconazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-1,2,4-triazole is used as an intermediate for the preparation of antifungal agent like fluconazole.

Solubility
Soluble in water and methanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-13-4
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.082
MDL Number MFCD00003099
InChI Key FMCUPJKTGNBGEC-UHFFFAOYSA-N
Synonym 4-amino-1,2,4-triazole, 4-amino-4h-1,2,4-triazole, 4h-1,2,4-triazol-4-amine, 1-amino-1,3,4-triazole, 4h-1,2,4-triazole, 4-amino, 1-amino-1h-1,3,4-triazole, 4-amino-1,2,4 4h-triazole, 4h-1,2,4-triazol-4-ylamine, 1-amino-1,4-triazole, 4-amino-1,4-triazole
PubChem CID 11432
IUPAC Name 1,2,4-triazol-4-amine
SMILES C1=NN=CN1N

Specifications

Melting Point 83°C to 88°C
Quantity 100 g
Beilstein 107563
Solubility Information Soluble in water and methanol.
Formula Weight 84.08
Percent Purity 99%
Chemical Name or Material 4-Amino-1,2,4-triazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.