missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-(2-pyridyl)piperidine hydrochloride, 97%

Product Code. 15459145
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459145 250 mg 250mg
15407997 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459145 Supplier Thermo Scientific Alfa Aesar Supplier No. H51108.MD
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 77145-39-2 | C10H16ClN3 | 213.709 g/mol

4-Amino-1-(2-pyridyl)piperidine hydrochloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-1-(2-pyridyl)piperidine hydrochloride is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77145-39-2
Molecular Formula C10H16ClN3
Molecular Weight (g/mol) 213.709
MDL Number MFCD09607833
InChI Key HNICPQFPDAEZKW-UHFFFAOYSA-N
Synonym 1-pyridin-2-yl piperidin-4-amine hydrochloride, 4-amino-1-2-pyridyl piperidine hydrochloride, 1-2-pyridyl-4-piperidylamine, chloride, 1-2-pyridyl-4-piperidinamine hydrochloride, 1-pyridin-2-yl piperidin-4-amine-hydrogen chloride 1/1, 1-pyridin-2-yl-piperazine hydrochloride; 1-2-pyridyl-4-piperidinamine hydrochloride
PubChem CID 45786931
IUPAC Name 1-pyridin-2-ylpiperidin-4-amine;hydrochloride
SMILES C1CN(CCC1N)C2=CC=CC=N2.Cl

Specifications

Quantity 250 mg
Formula Weight 213.71
Percent Purity 97%
Chemical Name or Material 4-Amino-1-(2-pyridyl)piperidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.