missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-Boc-piperidine, 97%

Product Code. 15453987
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453987 5 g 5g
15443987 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453987 Supplier Thermo Scientific Alfa Aesar Supplier No. H30779.06
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 87120-72-7 | C10H21N2O2 | 201.29 g/mol

4-Amino-1-Boc-piperidine is a chemical reagent used in the preparation of pharmaceutical compounds. Used in the synthesis of bromodomain inhibitors as well as HepG2 cell cycle inhibitors used in anti-tumor therapy. Also employed in a microwave-assisted solid-phase synthesis of N-substituted piperidines via direct annulation of primary amines with resin-bound dimesylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-1-Boc-piperidine is a chemical reagent used in the preparation of pharmaceutical compounds. Used in the synthesis of bromodomain inhibitors as well as HepG2 cell cycle inhibitors used in anti-tumor therapy. Also employed in a microwave-assisted solid-phase synthesis of N-substituted piperidines via direct annulation of primary amines with resin-bound dimesylates.

Solubility
Insoluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87120-72-7
Molecular Formula C10H21N2O2
Molecular Weight (g/mol) 201.29
MDL Number MFCD01076201
InChI Key LZRDHSFPLUWYAX-UHFFFAOYSA-O
Synonym 4-amino-1-boc-piperidine, 1-boc-4-aminopiperidine, n-boc-4-aminopiperidine, n-boc-4-amino-piperidine, 4-amino-1-n-boc-piperidine, 4-amino-n-boc piperidine, 1-boc-4-amino piperidine, 1-n-boc-4-aminopiperidine, 1-t-butoxycarbonyl-4-aminopiperidine, n-1-boc-4-aminopiperidine
PubChem CID 1268291
IUPAC Name tert-butyl 4-aminopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC([NH3+])CC1

Specifications

Melting Point 44°C to 48°C
Quantity 5 g
UN Number UN3259
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material 4-Amino-1-Boc-piperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.