missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,2-difluoro-1,3-benzodioxole, 97+%

Product Code. 11419837
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11419837 Supplier Thermo Scientific Alfa Aesar Supplier No. B22797.06
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106876-54-4 | C7H5F2NO2 | 173.12 g/mol

4-Amino-2,2-difluoro-1,3-benzodioxole is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-2,2-difluoro-1,3-benzodioxole is used in organic synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106876-54-4
Molecular Formula C7H5F2NO2
Molecular Weight (g/mol) 173.12
MDL Number MFCD00792416
InChI Key RXIKYXZKSIARLN-UHFFFAOYSA-N
Synonym 4-amino-2,2-difluoro-1,3-benzodioxole, 2,2-difluorobenzo d 1,3 dioxol-4-amine, 2,3-difluoromethylenedioxy aniline, 2,2-difluoro-4-amino-1,3-benzodioxole, 1,3-benzodioxol-4-amine, 2,2-difluoro, 2,2-difluoro-2h-1,3-benzodioxol-4-amine, 2,2-difluoro-benzo 1,3 dioxol-4-ylamine, 2,3-difluoromethylenedioxy-aniline, acmc-1btze, 2,2-difluoro-1,3-benzodioxole-4-amine
PubChem CID 2736891
IUPAC Name 2,2-difluoro-2H-1,3-benzodioxol-4-amine
SMILES NC1=CC=CC2=C1OC(F)(F)O2

Specifications

Quantity 5 g
UN Number UN2810
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 173.12
Percent Purity ≥97%
Chemical Name or Material 4-Amino-2,2-difluoro-1,3-benzodioxole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.