missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,6-dimethylpyridine, 98%

Product Code. p-7054080
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15498558 250 mg 250mg
15478558 1 g 1g
15488558 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15498558 Supplier Thermo Scientific Alfa Aesar Supplier No. H61970.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3512-80-9 | C7H10N2 | 122.171 g/mol

4-Amino-2,6-dimethylpyridine, is an important raw material and intermediate used in pharmaceuticals, agrochemicals, dyestuff, field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-2,6-dimethylpyridine, is an important raw material and intermediate used in pharmaceuticals, agrochemicals, dyestuff, field.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3512-80-9
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00130078
InChI Key ZJXMKPARTVOUAM-UHFFFAOYSA-N
PubChem CID 77047
IUPAC Name 2,6-dimethylpyridin-4-amine
SMILES CC1=CC(=CC(=N1)C)N

Specifications

Quantity 250 mg
UN Number UN2811
Synonym 4-amino-2,6-dimethylpyridine, 2,6-dimethyl-pyridin-4-ylamine, 2,6-dimethylpyridin-4-ylamine, 4-amino-2,6-lutidine, 4-pyridinamine, 2,6-dimethyl, 2,6-dimethylpyridin-4yl-amine, 2,6-dimethyl-4-pyridinamine, 2,6-dimethyl-4-aminopyridine, pubchem19474, 4-amino-2, 6-lutidine
Solubility Information Slightly soluble in water.
Formula Weight 122.17
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Amino-2,6-dimethylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.