missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Amino-2-chloro-5-fluoropyrimidine, 97%

Product Code. 15452108
Change view
Click to view available options
Quantity:
1g
Unit Size:
1g
Product Code. Quantity unitSize
15452108 1g 1g
1 options
This item is not returnable. View return policy

Product Code. 15452108

Brand: Alfa Aesar™ H55765.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 155-10-2
Molecular Formula C4H3ClFN3
MDL Number MFCD00057344
Quantity 1g
Synonym 4-amino-2-chloro-5-fluoropyrimidine, 2-chloro-5-fluoro-pyrimidin-4-ylamine, 2-chloro-4-amino-5-fluoropyrimidine, 2-chloro-5-fluoro-4-aminopyrimidine, 2-chloro-5-fluoropyrimidin-4-ylamine, 2-chloro-5-fluoropyrimidine-4-ylamine, 4-pyrimidinamine, 2-chloro-5-fluoro, 4-pyrimidinamine, 2-chloro-5-fluoro-9ci, pubchem6930, 2-chlor-4-amino-5-fluorpyrimidin
InChI Key SLQAJWTZUXJPNY-UHFFFAOYSA-N
SMILES C1=C(C(=NC(=N1)Cl)N)F
IUPAC Name 2-chloro-5-fluoropyrimidin-4-amine
Molecular Weight (g/mol) 147.537
PubChem CID 254372
Formula Weight 147.54
Percent Purity 97%
Chemical Name or Material 4-Amino-2-chloro-5-fluoropyrimidine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.