missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chloro-5-fluoropyrimidine, 98%

Product Code. 10356254 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10356254 5 g Glass bottle 5g
10183334 250 mg Glass bottle 250mg
10244864 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10356254 Supplier Thermo Scientific Acros Supplier No. 399980050
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 155-10-2 | C4H3ClFN3 | 147.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155-10-2
Molecular Formula C4H3ClFN3
Molecular Weight (g/mol) 147.54
MDL Number MFCD00057344
InChI Key SLQAJWTZUXJPNY-UHFFFAOYSA-N
Synonym 4-amino-2-chloro-5-fluoropyrimidine, 2-chloro-5-fluoro-pyrimidin-4-ylamine, 2-chloro-4-amino-5-fluoropyrimidine, 2-chloro-5-fluoro-4-aminopyrimidine, 2-chloro-5-fluoropyrimidin-4-ylamine, 2-chloro-5-fluoropyrimidine-4-ylamine, 4-pyrimidinamine, 2-chloro-5-fluoro, 4-pyrimidinamine, 2-chloro-5-fluoro-9ci, pubchem6930, 2-chlor-4-amino-5-fluorpyrimidin
PubChem CID 254372
IUPAC Name 2-chloro-5-fluoropyrimidin-4-amine
SMILES C1=C(C(=NC(=N1)Cl)N)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 193°C to 195°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 147.54
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-chloro-5-fluoropyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Amino-2-chloro-5-fluoropyrimidine
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.