missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chloro-6,7-dimethoxyquinazoline, TRC

Product Code. 30011887
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011887 25 g 25g
30006957 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011887 Supplier Toronto Research Chemicals Supplier No. TRCA60343925G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23680-84-4
Molecular Formula C10 H10 Cl N3 O2
Molecular Weight (g/mol) 239.66
Synonym 2-Chloro-6,7-dimethoxyquinazolin-4-amine, Doxazosin Mesilate Imp. F (EP), Terazosin Hydrochloride Imp. A (EP), Alfuzosin Hydrochloride Imp. B (EP), Prazosin Hydrochloride Imp. A (EP), Quinazoline, 4-amino-2-chloro-6,7-dimethoxy- (8CI), 2-Chloro-4-amino-6,7-dimethoxyquinazoline, 2-Chloro-6,7-dimethoxy-4-quinazolinamine, 4-Amino-2-chloro-6,7-dimethoxyquinazoline
IUPAC Name 2-chloro-6,7-dimethoxyquinazolin-4-amine
SMILES COc1cc2nc(Cl)nc(N)c2cc1OC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H10ClN3O2/c1-15-7-3-5-6(4-8(7)16-2)13-10(11)14-9(5)12/h3-4H,1-2H3,(H2,12,13,14)
Formula Weight 239.0462
Chemical Name or Material 2-Chloro-6,7-dimethoxyquinazolin-4-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.