missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chloropyridine, 98%

Product Code. 11409607
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409607 1 g 1g
11419607 5 g 5g
11399429 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409607 Supplier Thermo Scientific Alfa Aesar Supplier No. B22098.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14432-12-3 | C5H5ClN2 | 128.559 g/mol

2-Chloropyridin-4-amine (14432-12-3) is used as a medicine, pigment intermediates. It is an additive of plant growth regulator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14432-12-3
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD00060089
InChI Key BLBDTBCGPHPIJK-UHFFFAOYSA-N
Synonym 4-amino-2-chloropyridine, 2-chloro-4-aminopyridine, 2-chloro-pyridin-4-ylamine, 4-pyridinamine, 2-chloro, 2-chloro-4-pyridinamine, 2-chlorpyridin-4-amin, 4-amino-6-chloropyridine, 2-chloro-4-pyridylamine, 2-chloro-4-amino pyridine, 4-amino-2-chloro pyridine
PubChem CID 84432
IUPAC Name 2-chloropyridin-4-amine
SMILES C1=CN=C(C=C1N)Cl

Specifications

Melting Point 91°C to 94°C
Boiling Point 153°C (5 mmHg)
Packaging Plastic bottle
Quantity 1 g
Beilstein 108671
Solubility Information Slightly soluble in water.
Formula Weight 128.56
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-chloropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.