missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chloropyrimidine, 98%

Product Code. 15492258
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492258 5 g 5g
15402268 250 mg 250mg
15482258 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492258 Supplier Thermo Scientific Alfa Aesar Supplier No. H55875.06
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7461-50-9 | C4H4ClN3 | 129.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7461-50-9
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.55
MDL Number MFCD00194051
InChI Key LPBDZVNGCNTELM-UHFFFAOYSA-N
Synonym 4-amino-2-chloropyrimidine, 2-chloro-4-aminopyrimidine, 2-chloro-4-pyrimidinamine, 2-chloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 2-chloro, 2-chloropyrimidin-4-ylamine, 2-chloro-4-amino-pyrimidine, 2-chloropyrimidine-4-ylamine, zlchem 116, pubchem6955
PubChem CID 345752
IUPAC Name 2-chloropyrimidin-4-amine
SMILES NC1=CC=NC(Cl)=N1

Specifications

Melting Point 197°C to 205°C
Quantity 5 g
Formula Weight 129.55
Percent Purity 98%
Chemical Name or Material 4-Amino-2-chloropyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.