missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-ethoxy-5-nitrobenzoic Acid Methyl Ester, TRC

Product Code. 30324173
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324173 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324173 Supplier Toronto Research Chemicals Supplier No. TRCA6089851G
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Amino-2-ethoxy-5-nitrobenzoic Acid Methyl Ester
CAS 86718-17-4
Molecular Formula C10H12N2O5
Quantity 1 g
Synonym Methyl 4-Amino-2-ethoxy-5-nitrobenzoate
InChI Formula InChI=1S/C10H12N2O5/c1-3-17-9-5-7(11)8(12(14)15)4-6(9)10(13)16-2/h4-5H,3,11H2,1-2H3
SMILES CCOc1cc(N)c(cc1C(=O)OC)[N+](=O)[O-]
IUPAC Name methyl 4-amino-2-ethoxy-5-nitrobenzoate
Molecular Weight (g/mol) 240.21
Formula Weight 240.0746
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.