missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Product Code. p-7040209
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15405356 250 mg 250mg
15495346 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15405356 Supplier Thermo Scientific Alfa Aesar Supplier No. H59916.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 819057-45-9
Molecular Formula C12H17BFNO2
Molecular Weight (g/mol) 237.08
MDL Number MFCD09951877
InChI Key FLMNWVXAEGUVNY-UHFFFAOYSA-N
Synonym 4-amino-2-fluorophenylboronic acid pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-2-fluorobenzeneboronic acid pinacol ester, benzenamine, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-amino-2-fluorophenyl boronic acid pinacol ester, 4-amino-2-fluorophenylboronic acid, pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl phenylamine, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline
PubChem CID 44755207
IUPAC Name 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=C(F)C=C(N)C=C1

Specifications

Quantity 250 mg
Formula Weight 237.08
Percent Purity 96%
Chemical Name or Material 4-Amino-2-fluorobenzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.