missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2-fluorophenol, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 4-Amino-2-fluorophenol |
| CAS | 399-96-2 |
| Molecular Formula | C6H6FNO |
| Quantity | 5 g |
| Synonym | 3-Fluoro-4-hydroxyaniline, 2-Fluoro-4-aminophenol |
| InChI Formula | InChI=1S/C6H6FNO/c7-5-3-4(8)1-2-6(5)9/h1-3,9H,8H2 |
| SMILES | Nc1ccc(O)c(F)c1 |
| IUPAC Name | 4-amino-2-fluorophenol |
| Molecular Weight (g/mol) | 127.12 |
| Formula Weight | 127.0433 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction