missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-isopropyl-5-methylphenol hydrochloride, 97%

Product Code. 11302977
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302977 1 g 1g
11312977 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302977 Supplier Thermo Scientific Alfa Aesar Supplier No. L16282.03
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 6321-11-5 | C6H10O6 | 178.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6321-11-5
MDL Number MFCD00035504

Specifications

Melting Point 258°C to 262°C
Odor Odorless
Molecular Formula C10H15ClNO
Quantity 1 g
Beilstein 3700651
Synonym 4-amino-2-isopropyl-5-methylphenol hydrochloride, 6-aminothymol hydrochloride, phenol, 4-amino-5-methyl-2-1-methylethyl-, hydrochloride, 4-amino-2-isopropyl-5-methyl-phenolhydrochloride, 4-amino-2-isopropyl-5-methyl-phenol hydrochloride, 4-amino-5-methyl-2-propan-2-yl phenol hydrochloride, phenol, 4-amino-5-methyl-2-1-methylethyl-, hydrochloride 1:1, 6-aminothymol hcl, 6-aminothymolhydrochlorid, 6-aminothymol hydrovhloride
InChI Key YLWORLAQZBRLSS-UHFFFAOYSA-N
SMILES CC1=C(C=C(C(=C1)O)C(C)C)N.Cl
IUPAC Name 4-amino-5-methyl-2-propan-2-ylphenol;hydrochloride
Molecular Weight (g/mol) 200.69
PubChem CID 80603
Formula Weight 201.7
Percent Purity 97%
Chemical Name or Material 4-Amino-2-isopropyl-5-methylphenol hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.