missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-(methylthio)pyrimidine-5-carbonitrile, 97%, Thermo Scientific™

Product Code. 10150412 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10150412 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10150412 Supplier Thermo Scientific Acros Supplier No. 354310010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 770-30-9
CAS Min % 96.0
CAS Max % 100.0
Melting Point 240°C to 242°C
Color Cream to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H6N4S
MDL Number MFCD00023237
Quantity 1 g
Beilstein 25, V,17, 157
Synonym 4-amino-2-methylthio pyrimidine-5-carbonitrile, 4-amino-2-methylsulfanyl pyrimidine-5-carbonitrile, 4-amino-2-methylthio-5-pyrimidinecarbonitrile, 4-amino-2-methylsulfanyl-pyrimidine-5-carbonitrile, 4-amino-2-methylthiopyrimidine-5-carbonitrile, maybridge1_004789, acmc-20a4j5, 4-amino-5-cyano-2-methylthiopyrimidine, 4-amino-5-cyano-2-methylthio pyrimidine
InChI Key VYDHBDPZNLVELI-UHFFFAOYSA-N
SMILES CSC1=NC=C(C#N)C(N)=N1
IUPAC Name 4-amino-2-(methylsulfanyl)pyrimidine-5-carbonitrile
Molecular Weight (g/mol) 166.20
PubChem CID 295757
Formula Weight 166.21
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Amino-2-(methylthio)pyrimidine-5-carbonitrile, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Amino-2-(methylthio)pyrimidine-5-carbonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.