missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole, 99+%, Thermo Scientific Chemicals

Product Code. 11384404
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384404 5 g 5g
11394404 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384404 Supplier Thermo Scientific Alfa Aesar Supplier No. L00982.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole is a reagent used in the determination of aldehydes and other reactive chemicals.was used in purpald assay for determination of bacterial polysaccharides containing substituted or unsubstituted glycol in residues such as glycerol, ribitol, arabinitol, furanosyl galactose and sialic acid. It was used as chromogen during the determination of catalase activity in small tissue samples. It was used as reagent in determination of formaldehyde. It was used for derivatization of neutral monosaccharides during HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole is a reagent used in the determination of aldehydes and other reactive chemicals.was used in purpald assay for determination of bacterial polysaccharides containing substituted or unsubstituted glycol in residues such as glycerol, ribitol, arabinitol, furanosyl galactose and sialic acid. It was used as chromogen during the determination of catalase activity in small tissue samples. It was used as reagent in determination of formaldehyde. It was used for derivatization of neutral monosaccharides during HPLC.

Solubility
Soluble in Dimethyl sulfoxide. (DMSO)

Notes
Store at -20°C. Avoid contact with heat. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1750-12-5
Molecular Formula C2H6N6S
Molecular Weight (g/mol) 146.172
MDL Number MFCD00003098
InChI Key RDIMQHBOTMWMJA-UHFFFAOYSA-N
Synonym 4-amino-3-hydrazino-5-mercapto-1,2,4-triazole, purpald, 4-amino-3-hydrazino-1,2,4-triazol-5-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, hydrazone, purpald r, 4-amino-5-hydrazino-1,2,4-triazole-3-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, 3-hydrazone, 4-amino-5-hydrazinyl-4h-1,2,4-triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 4-amino-5-hydrazinyl-2,4-dihydro, 4-amino-5-hydrazinyl-1,2,4-triazole-3-thiol
PubChem CID 2723946
IUPAC Name 4-amino-3-hydrazinyl-1H-1,2,4-triazole-5-thione
SMILES C1(=S)NN=C(N1N)NN

Specifications

Melting Point ∽220°C (decomposition)
Quantity 5 g
UN Number UN1325
Beilstein 134867
Solubility Information Soluble in Dimethyl sulfoxide. (DMSO)
Formula Weight 146.17
Percent Purity ≥99%
Chemical Name or Material 4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole
compliance-icons compliance-icons
Product Identifier
  • 4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.