missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-Hydroxybutanoic Acid, TRC

Product Code. 30323633
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323633 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323633 Supplier Toronto Research Chemicals Supplier No. TRCA6112701G
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Amino-3-Hydroxybutanoic Acid
CAS 924-49-2
Molecular Formula C4H9NO3
Quantity 1 g
Synonym (±)-4-Amino-3-hydroxy-butanoic Acid, DL-4-Amino-3-hydroxybutyric Acid, γ-Amino-β-hydroxybutyric Acid, (RS)-3-Hydroxy-γ-aminobutyric Acid, (RS)-4-Amino-3-hydroxybutanoic Acid, (RS)-γ-Amino-β-hydroxybutyric Acid, (±)-3-Hydroxy-4-aminobutanoic Acid, (±)-4-Amino-3-hydroxybutanoic Acid, (±)-4-Amino-3-hydroxybutyric Acid, (±)-β-Hydroxy-GABA, 3-Hydroxy-4-aminobutanoic Acid, 3-Hydroxy-4-aminobutyric Acid, 3-Hydroxy-GABA, 4-Amino-3-hydroxybutanoic Acid, 4-Amino-3-hydroxybutyric Acid, Bogil, Buksamin, Buxamine, DL-3-Hydroxy-4-aminobutyric Acid, DL-4-Amino-3-hydroxybutanoic Acid, DL-4-Amino-3-hydroxybutyric Acid, DL-β-Hydroxy-γ-aminobutyric Acid, DL-γ-Amino-β-hydroxybutyric Acid, GABOB, Gabobe, Gabomade, Gaboril, Gamibetal, Gaminal, Idramina, NSC 40244, β-Hydroxy-GABA, β-Hydroxy-γ-aminobutyric Acid, γ-Amino-β-hydroxybutyric Acid
InChI Formula InChI=1S/C4H9NO3/c5-2-3(6)1-4(7)8/h3,6H,1-2,5H2,(H,7,8)
SMILES OC(CC(O)CN)=O
IUPAC Name 4-amino-3-hydroxybutanoic acid
Molecular Weight (g/mol) 119.12
Formula Weight 119.0582
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.