missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-methylbenzoic Acid, TRC

Product Code. 30310418
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30310418 5 g 5g
30326706 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30310418 Supplier Toronto Research Chemicals Supplier No. TRCA6145855G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Amino-3-methylbenzoic Acid
CAS 2486-70-6
Molecular Formula C8 H9 N O2
Quantity 5 g
Synonym 4-Amino-3-methylbenzoic Acid, m-Toluic acid, 4-amino- (6CI,7CI,8CI), 4-Amino-3-methylbenzoic acid, 3-Methyl-4-aminobenzoic acid, 4-Amino-5-methylbenzoic acid, BTS 28369, NSC 227945
InChI Formula InChI=1S/C8H9NO2/c1-5-4-6(8(10)11)2-3-7(5)9/h2-4H,9H2,1H3,(H,10,11)
SMILES Cc1cc(ccc1N)C(=O)O
IUPAC Name 4-amino-3-methylbenzoic acid
Molecular Weight (g/mol) 151.162
Formula Weight 151.0633
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.