missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-3-methylbenzoic Acid, TRC
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | 4-Amino-3-methylbenzoic Acid |
| CAS | 2486-70-6 |
| Molecular Formula | C8 H9 N O2 |
| Quantity | 1 g |
| Synonym | 4-Amino-3-methylbenzoic Acid, m-Toluic acid, 4-amino- (6CI,7CI,8CI), 4-Amino-3-methylbenzoic acid, 3-Methyl-4-aminobenzoic acid, 4-Amino-5-methylbenzoic acid, BTS 28369, NSC 227945 |
| InChI Formula | InChI=1S/C8H9NO2/c1-5-4-6(8(10)11)2-3-7(5)9/h2-4H,9H2,1H3,(H,10,11) |
| SMILES | Cc1cc(ccc1N)C(=O)O |
| IUPAC Name | 4-amino-3-methylbenzoic acid |
| Molecular Weight (g/mol) | 151.162 |
| Formula Weight | 151.0633 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction