missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, 99+%, Certified AR for Analysis, Fisher Chemical™

Product Code. 10795711 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
Packaging:
HDPE plastic bottle
Unit Size:
1 L
2.5 L
5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10795711 2.5 L HDPE plastic bottle 2.5 L
10579570 500 mL HDPE plastic bottle 500mL
12656707 1 L HDPE plastic bottle 1 L
12666707 5 L HDPE plastic bottle 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10795711 Supplier Fisher Chemical Supplier No. G/0650/17
Spotlight Offers
Use promo code 28036
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

C3H8O3, CAS Number-56-81-5, propanetriol, trihydroxypropane, glycyl alcohol, glycerol, glyceritol, glycerin, glycerine, osmoglyn, 1,2,3-propanetriol, 1,2,3-trihydroxypropane, 2.5L, 290 deg.C, CHEBI:17754, Undesignated, 92.09g/mol, PEDCQBHIVMGVHV-UHFFFAOYSA-N, propane-1,2,3-triol, 18 deg.C, 4722 | CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

C3H8O3, CAS Number-56-81-5, glycerol, propanetriol, glycyl alcohol, glycerin, 1,2,3-trihydroxypropane, glycerine, 1,2,3-propanetriol, trihydroxypropane, osmoglyn, glyceritol, 1L, 290 deg.C, CHEBI:17754, Undesignated, 92.09g/mol, PEDCQBHIVMGVHV-UHFFFAOYSA-N, propane-1,2,3-triol, 18 deg.C, 4722

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
pH 5
Boiling Point 290°C
Packaging HDPE plastic bottle
Quantity 2.5 L
Vapor Density 3.17
Formula Weight 92.09g/mol
Vapor Pressure 0.003mbar 50
Physical Form Very Viscous Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.