missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-penten-2-one, 96%

Product Code. 11318125
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11318125 1 g 1g
11328125 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11318125

Brand: Thermo Scientific Alfa Aesar L08379.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1118-66-7 | C5H9NO | 99.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-66-7
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00043715
InChI Key OSLAYKKXCYSJSF-ONEGZZNKSA-N
Synonym acetylacetonamine, fluoral-p, 4-amino-3-penten-2-one, e-4-aminopent-3-en-2-one, 3-penten-2-one, 4-amino, 3e-4-aminopent-3-en-2-one, 4-aminopent-3-en-2-one, 4-amino-pent-3-en-2-one, e-4-amino-3-pentene-2-one, 2-acetyl-1-amino-1-methylethylene
PubChem CID 5367854
ChEBI CHEBI:51695
IUPAC Name (E)-4-aminopent-3-en-2-one
SMILES CC(=CC(=O)C)N

Specifications

Melting Point 36°C to 41°C
Boiling Point 209°C to 213°C
Quantity 1 g
Beilstein 1209344
Formula Weight 99.13
Percent Purity 96%
Chemical Name or Material 4-Amino-3-penten-2-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.