missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-bromo-2-chloropyrimidine, 95%

Product Code. 15407978
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15407978 5 g 5g
15497968 1 g 1g
15417978 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15407978 Supplier Thermo Scientific Alfa Aesar Supplier No. H61429.06
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 205672-25-9 | C4H3BrClN3 | 208.44 g/mol

4-Amino-5-bromo-2-chloropyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-5-bromo-2-chloropyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Storage temperature: 2-4°C. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205672-25-9
Molecular Formula C4H3BrClN3
Molecular Weight (g/mol) 208.44
MDL Number MFCD07782040
InChI Key QOWALNIZDHZTSM-UHFFFAOYSA-N
Synonym 4-amino-5-bromo-2-chloropyrimidine, 4-pyrimidinamine, 5-bromo-2-chloro, 5-bromo-2-chloro-4-pyrimidinamine, 5-bromo-2-chloro-pyrimidin-4-ylamine, 5-bromo-2-chloropyrimidine-4-ylamine, pubchem18570, acmc-209fbz, 5-bromo-2-chloro-pyrimidin-4-amine, 5-bromo-2-chloropyrimidin-4-ylamine
PubChem CID 2763293
IUPAC Name 5-bromo-2-chloropyrimidin-4-amine
SMILES NC1=NC(Cl)=NC=C1Br

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 208.44
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Amino-5-bromo-2-chloropyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.