missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-mercapto-1H-pyrazolo[3,4-d]pyrimidine, 94%, Thermo Scientific™

Product Code. 11387056
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11387056 250 mg 250mg
11377056 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11387056

Brand: Thermo Scientific Alfa Aesar L06934.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23771-52-0
Molecular Formula C5H5N5S
Molecular Weight (g/mol) 167.19
MDL Number MFCD00005690
InChI Key YJMNLDSYAAJOPX-UHFFFAOYSA-N
Synonym 4-amino-6-mercaptopyrazolo 3,4-d pyrimidine, 4-amino-1h-pyrazolo 3,4-d pyrimidine-6-thiol, 4-aminopyrazolo 5,4-d pyrimidine-6-thiol, 4lkq, acmc-20ampb, 4-amino-6-mercaptopyrazolo 3?4-d pyrimidine, 4-amino-6-mercpato pyrazolo 3,4-d pyrimidine, 4-amino-2h-pyrazolo 3,4-d pyrimidine-6-thiol, 4-amino-1h-pyrazolo-3,4-d-pyrimidin-6-thiol
PubChem CID 1268262
IUPAC Name 4-amino-1H,2H,6H-pyrazolo[3,4-d]pyrimidine-6-thione
SMILES NC1=NC(=S)N=C2NNC=C12

Specifications

Melting Point >300°C
Quantity 250 mg
Beilstein 152193
Formula Weight 167.19
Percent Purity 94%
Chemical Name or Material 4-Amino-6-mercapto-1H-pyrazolo[3,4-d]pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.