missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-N-[2-(4-aminophenoxy)ethyl]-N-methylbenzeneethanamine, TRC

Product Code. 30325773
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325773 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325773 Supplier Toronto Research Chemicals Supplier No. TRCA581980100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Amino-N-[2-(4-aminophenoxy)ethyl]-N-methylbenzeneethanamine
CAS 115256-13-8
Molecular Formula C17H23N3O
Quantity 100 mg
Synonym 4-Amino-N-[2-(4-aminophenoxy)ethyl]-N-methylbenzeneethanamine
InChI Formula InChI=1S/C17H23N3O/c1-20(11-10-14-2-4-15(18)5-3-14)12-13-21-17-8-6-16(19)7-9-17/h2-9H,10-13,18-19H2,1H3
SMILES CN(CCC1=CC=C(C=C1)N)CCOC2=CC=C(C=C2)N
IUPAC Name 4-[2-[2-(4-aminophenoxy)ethyl-methylamino]ethyl]aniline
Molecular Weight (g/mol) 285.38
Formula Weight 285.1841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.