missing translation for 'onlineSavingsMsg'
Learn More

4'-Aminoacetophenone, 99%

Product Code. 11490723
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
1000 g
Unit Size:
1000g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490723 50 g 50g
11369429 25 g 25g
11400733 250 g 250g
11480723 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11490723 Supplier Thermo Scientific Alfa Aesar Supplier No. A13213.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99-92-3 | C8H9NO | 135.17 g/mol

4'-Aminoacetophenone, is used in pharmaceuticals, medicines and other organic products. It is also used as a reagent for the photometric determination of Ce. As pharmaceutical intemediate or in the determination of palladium and vitaminB1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Aminoacetophenone, is used in pharmaceuticals, medicines and other organic products. It is also used as a reagent for the photometric determination of Ce. As pharmaceutical intemediate or in the determination of palladium and vitaminB1.

Solubility
It is soluble in hot water, ethanol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. acids, Incompatible to Acid chlorides, Acid anhydrides, Chloroformates, Strong oxidizing agents, Strong reducing agents. Stable under ordinary conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-92-3
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00007896
InChI Key GPRYKVSEZCQIHD-UHFFFAOYSA-N
Synonym 4'-aminoacetophenone, 4-aminoacetophenone, 1-4-aminophenyl ethanone, p-aminoacetophenone, 4-acetylaniline, p-acetylaniline, ethanone, 1-4-aminophenyl, p-aminoacetylbenzene, 1-4-aminophenyl ethan-1-one, acetophenone, p-amino
PubChem CID 7468
IUPAC Name 1-(4-aminophenyl)ethanone
SMILES CC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 103°C to 108°C
Boiling Point 292°C to 294°C
Quantity 50 g
Beilstein 471493
Merck Index 14,413
Solubility Information It is soluble in hot water,ethanol and ether.
Formula Weight 135.17
Percent Purity 99%
Chemical Name or Material 4'-Aminoacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.