missing translation for 'onlineSavingsMsg'
Learn More

4'-Aminoacetophenone, 99%

Product Code. 10040710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10040710 10 g Glass bottle 10g
10172890 25 g Glass bottle 25g
10764971 100 g Glass bottle 100g
10321294 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10040710 Supplier Thermo Scientific Acros Supplier No. 103090100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 99-92-3 | C8H9NO | 135.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-92-3
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00007896
InChI Key GPRYKVSEZCQIHD-UHFFFAOYSA-N
Synonym 4'-aminoacetophenone, 4-aminoacetophenone, 1-4-aminophenyl ethanone, p-aminoacetophenone, 4-acetylaniline, p-acetylaniline, ethanone, 1-4-aminophenyl, p-aminoacetylbenzene, 1-4-aminophenyl ethan-1-one, acetophenone, p-amino
PubChem CID 7468
IUPAC Name 1-(4-aminophenyl)ethanone
SMILES CC(=O)C1=CC=C(N)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 96.0°C to 102.0°C
Color Brown to Yellow
Boiling Point 293.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NC6H4COCH3
Quantity 10 g
Beilstein 14, 46
Fieser 04,18
Merck Index 14, 413
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in hcl,soluble in alcohol and ether,sparingly soluble in benzene
Formula Weight 135.17
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4'-Aminoacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Aminoacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.