missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoantipyrine, 98%

Product Code. 10795161 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10795161 25 g Glass bottle 25g
10497660 100 g Glass bottle 100g
10705171 500 g Glass bottle 500g
11946863 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10795161 Supplier Thermo Scientific Acros Supplier No. 103150250
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

4-Aminoantipyrine, 98%, C11H13N3O, CAS Number-83-07-8, 4-aminoantipyrene, aminoantipyrin, ampyrone, 4-aminoantipyrine, 4-aminophenazone, metapirazone, solnapyrin-a, aminoazophene, solvapyrin-a, aminoantipyrine, 25g, (5% aq. soln.) clear, 24,273, 100.0, 97.5, CHEBI:59026, Brown-Yellow to Yellow | CAS: 83-07-8 | C11H13N3O | 203.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-07-8
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.25
MDL Number MFCD00003145
InChI Key RLFWWDJHLFCNIJ-UHFFFAOYSA-N
Synonym 4-aminoantipyrine, ampyrone, metapirazone, aminoantipyrine, solvapyrin-a, aminoazophene, 4-aminophenazone, 4-aminoantipyrene, aminoantipyrin, solnapyrin-a
PubChem CID 2151
ChEBI CHEBI:59026
IUPAC Name 4-amino-1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 105.5°C to 110.0°C
Color Brown-Yellow to Yellow
Density 0.8000g/mL
Loss on Drying 1.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 24,273
Merck Index 15,413
Specific Gravity 0.8
Solubility Information (5% in water) Clear
Formula Weight 203.25
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 4-Aminoantipyrine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Aminoantipyrine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.