missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminoazobenzene-3,4'-disulfonic Acid Disodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Aminoazobenzene-3,4'-disulfonic Acid Disodium Salt |
| CAS | 2706-28-7 |
| Molecular Formula | C12 H9 N3 O6 S2 Na2 |
| Quantity | 500 mg |
| Synonym | Sodium 2-Amino-5-((4-sulfonatophenyl)diazenyl)benzenesulfonate, C.I. Acid Yellow 9, C.I. Acid Yellow 9, disodium Salt, Fast Yellow S, Acetyl Yellow G, Acid Yellow 9, Acid Yellow AT, Acid Yellow G, Acid Yellow G Kond, Acid Yellow Geigy, Acid yellow, Acilan Yellow Extra, Amacid Yellow RG, C Yellow 9, C.I. 13015, C.I. Food Yellow 2, Cilefa Yellow R, Disodium 4-aminoazobenzene-3,4'-disulfonate, E 105, E 105 (dye), Fast Yellow, Fast Yellow AB, Fast Yellow Extra, Fast Yellow Extra Specially Pure, Fast Yellow S Extra specially Pure, Fast Yellow SX Extra Specially Pure, Fast Yellow Y, Food Yellow 2, Golden Yellow RUAF, Hexacol Acid Yellow G, Kiton Yellow Extra, Lacquer Yellow T, Wool Yellow G, Yellow Fast, Yellow Acid |
| InChI Formula | InChI=1S/C12H11N3O6S2.2Na/c13-11-6-3-9(7-12(11)23(19,20)21)15-14-8-1-4-10(5-2-8)22(16,17)18;;/h1-7H,13H2,(H,16,17,18)(H,19,20,21);;/q;2*+1/p-2 |
| SMILES | [Na+].[Na+].Nc1ccc(cc1S(=O)(=O)[O-])N=Nc2ccc(cc2)S(=O)(=O)[O-] |
| IUPAC Name | disodium;2-amino-5-[(4-sulfonatophenyl)diazenyl]benzenesulfonate |
| Molecular Weight (g/mol) | 401.32 |
| Formula Weight | 400.9728 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?