missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoazobenzene, TRC

Product Code. 30322280
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30322280

Brand: Toronto Research Chemicals TRCA5801101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Aminoazobenzene
CAS 60-09-3
Molecular Formula C12 H11 N3
Quantity 1 g
Synonym Benzenamine, 4-(2-phenyldiazenyl)-, Benzenamine, 4-(phenylazo)- (9CI), C.I. Solvent Yellow 1 (7CI,8CI), 4-(2-Phenyldiazenyl)benzenamine, 4-(Phenylazo)aniline, 4-(Phenylazo)benzenamine, 4-(Phenyldiazenyl)aniline, 4-Aminoazobenzene, 4-Aminoazobenzol, Aniline yellow, Brasilazina Oil Yellow G, C.I. 11000, Cellitazol R, Ceres Yellow R, Fast Spirit Yellow, Fat Yellow AAB, NSC 2032, Oil Soluble Aniline Yellow, Oil Yellow AAB, Oil Yellow AB, Oil Yellow AN, Oil Yellow B, Organol Yellow 2A, Solvent Yellow 1, Solvent yellow, Somalia Yellow 2G, Sudan Yellow R, p-(Phenylazo)aniline, p-Aminoazobenzene, p-Aminoazobenzol, p-Aminodiphenylimide
InChI Formula InChI=1S/C12H11N3/c13-10-6-8-12(9-7-10)15-14-11-4-2-1-3-5-11/h1-9H,13H2
SMILES Nc1ccc(cc1)N=Nc2ccccc2
IUPAC Name 4-[(E)-phenyldiazenyl]aniline
Molecular Weight (g/mol) 197.24
Formula Weight 197.0953
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.