missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzonitrile, 98%

Product Code. 10337680 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 10337680

Brand: Thermo Scientific Acros 164470500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 873-74-5 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-74-5
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00007821
InChI Key YBAZINRZQSAIAY-UHFFFAOYSA-N
Synonym 4-cyanoaniline, p-aminobenzonitrile, p-cyanoaniline, benzonitrile, 4-amino, aniline, p-cyano, benzonitrile, p-amino, 1-amino-4-cyanobenzene, unii-pb8v69y76t, 4-amino benzonitrile, 4-amino-benzonitrile
PubChem CID 13396
IUPAC Name 4-aminobenzonitrile
SMILES C1=CC(=CC=C1C#N)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 83.0°C to 87.0°C
Color Beige to Orange-Yellow or White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4CN
Quantity 50 g
Beilstein 14, 425
Solubility Information Solubility in water: insoluble
Formula Weight 118.14
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-Aminobenzonitrile
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.