missing translation for 'onlineSavingsMsg'
Learn More

(4-Aminobenzoyl)hydrazide, 95%

Product Code. 10068621 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10068621 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10068621 Supplier Thermo Scientific Acros Supplier No. 103200050
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5351-17-7 | C7H9N3O | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5351-17-7
CAS Min % 94.0
CAS Max % 100.0
Melting Point 224.0°C to 228.0°C
Color Beige or White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C7H9N3O
Linear Formula H2NC6H4CONHNH2
MDL Number MFCD00007606
Quantity 5 g
Beilstein 14, I, 570
Synonym 4-aminobenzhydrazide, amben hydrazide, p-aminobenzhydrazide, 4-aminobenzoic acid hydrazide, aminostimil, 4-aminobenzoylhydrazine, p-aminobenzoic acid hydrazide, p-aminobenzoyl hydrazide, benzoic acid, 4-amino-, hydrazide, p-aminobenzoic hydrazide
InChI Key WPBZMCGPFHZRHJ-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C(=O)NN)N
IUPAC Name 4-aminobenzohydrazide
Molecular Weight (g/mol) 151.17
PubChem CID 21450
Formula Weight 151.17
Percent Purity 95%
Physical Form Crystals or Powder
Chemical Name or Material (4-Aminobenzoyl)hydrazide, 95%
Show More Show Less
compliance-icons
Product Identifier
  • (4-Aminobenzoyl)hydrazide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.