missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobutyraldehyde diethyl acetal, 95%

Product Code. 10700605 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10700605

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6346-09-4 | C8H19NO2 | 161.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6346-09-4
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.24
MDL Number MFCD00008227
InChI Key GFLPSABXBDCMCN-UHFFFAOYSA-N
IUPAC Name 4,4-diethoxybutan-1-amine
SMILES CCOC(CCCN)OCC

Specifications

Color Colorless
Density 0.9000g/mL
Boiling Point 196.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula H2N(CH2)3CH(OC2H5)2
Refractive Index 1.4270 to 1.4290
Quantity 100 g
Beilstein 04,319
Specific Gravity 0.9
Synonym 2-chloro-5-aminophenol, 4-chloro-3-hydroxyaniline, phenol, 5-amino-2-chloro, 5-amino-2-chloro-phenol, 3-amino-6-chlorophenol, pubchem21902, 3-hydroxy-4-chloroaniline, 4-chloro-3-hydroxy aniline, acmc-1b64d, ksc494c4r
Solubility Information Solubility in water: miscible.
PubChem CID 80676
Formula Weight 161.24
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Aminobutyraldehyde diethyl acetal
Show More Show Less
compliance-icons
Product Identifier
  • 4-Aminobutyraldehyde diethyl acetal
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.